missing translation for 'onlineSavingsMsg'
Learn More

Chicoric acid, MedChemExpress

Product Code. p-200096304
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16494837

Brand: Medchem Express HYN0457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chicoric acid (Cichoric acid), an orally active dicaffeyltartaric acid, induces reactive oxygen species (ROS) generation.

Chicoric acid inhibits cell viability and induces mitochondria-dependent apoptosis in 3T3-L1 preadipocytes through ROS-mediated PI3K/Akt and MAPK signaling pathways. Chicoric acid increases glucose uptake, improves insulin resistance, and attenuates glucosamine-induced inflammation. Chicoric acid has antidiabetic properties and antioxidant, anti-inflammatory effects.

Specifications

Chemical Name or Material Chicoric acid
CAS 6537-80-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H18O12
Quantity 50 mg
Synonym Cichoric acid Dicaffeoyltartaric acid
Solubility Information H2O : 100 mg/mL (210.81 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.11 mM; Need ultrasonic)
SMILES O=C(O)[C@H](OC(/C=C/C1=CC=C(O)C(O)=C1)=O)[C@@H](OC(/C=C/C2=CC=C(O)C(O)=C2)=O)C(O)=O
Molecular Weight (g/mol) 474.37
Formula Weight 474.37
Percent Purity 95.21%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.