Learn More
Chicken anti-Rabbit IgG (H+L) Secondary Antibody, TRITC (Pre-adsorbed), Novus Biologicals™
Product Code. 18252525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Product Code. Quantity unitSize
18252525 0.5 mg 0.5mg
1 options
Product Code. 18252525
Brand: Novus Biologicals NBP175270
Chicken Polyclonal Antibody
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunomicroscopy.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG (H+L)
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunomicroscopy
Classification Polyclonal
Concentration LYOPH
Conjugate TRITC
Dilution Flow Cytometry 1:20-1:2000, ELISA, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20-1:2000
Host Species Chicken
Immunogen Purified Rabbit IgG (H&L)
Purification Method Affinity purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins. This antibody has been pre-adsorbed against human and mouse IgG.
Reconstitution Rehydrate with 0.55 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Target Species Rabbit
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
For Research Use Only