Learn More
Thermo Scientific™ Chest Freezer Racks
Product Code. 12633115 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Boxes (2 in.)
Boxes (3 in.)
General Freezer Items
Plates
For Use With (Equipment):
12.7 cu. ft. Freezers
12.7, 17, 20 cu. ft. Freezers
12.7, 20 cu. ft. Freezers
13, 17, 20 cu. ft. Freezers
20 cu. ft. Freezers
3 cu. ft. Chest Freezers
3 cu. ft. Freezers
Unit Size:
Each
Product Code. Holds For Use With (Equipment) unitSize
12633115 Boxes (3 in.) 13, 17, 20 cu. ft. Freezers Each
11796479 General Freezer Items 12.7, 20 cu. ft. Freezers Each
13504940 General Freezer Items 20 cu. ft. Freezers Each
13584930 General Freezer Items 12.7 cu. ft. Freezers Each
12623115 Boxes (2 in.) 3 cu. ft. Chest Freezers Each
12693665 Boxes (3 in.) 3 cu. ft. Freezers Each
11209769 Boxes (2 in.) 13, 17, 20 cu. ft. Freezers Each
11800553 Plates 12.7, 17, 20 cu. ft. Freezers Each
8 options
Product Code. 12633115
Brand: Thermo Scientific™ 398187
Only null left
Add to basket
Add to basket
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Holds Boxes (3 in.)
Material Stainless Steel
Positions 8 Boxes/Rack
For Use With (Equipment) 13, 17, 20 cu. ft. Freezers
Freezer Compatibility (Capacity) 13, 17, 20 cu. ft.
Width (English) 5.5 in.
Height (English) 26.5 in.
Depth (English) 5.5 in.
Depth (Metric) 14 cm
Product Type Side Access Rack Tower
Height (Metric) 67.3 cm
Width (Metric) 14 cm