Learn More
Saint Gobain Life Sciences Chemware™ PFA Narrow Neck Graduated Bottle
Click to view available options
Capacity:
100 mL (3.4 oz.)
250 mL (8.5 oz.)
5000 mL (1.3 gal.)
Description
- Ultra-pure - the lowest metal ion content of all fluoropolymers
- Unbreakable
- Chemically inert
- Non-stick, easy to clean
- Use for storage in pharmaceutical and semiconductor processing (intermediate stages)
Specifications
Capacity | 100 mL (3.4 oz.) |
Product Type | Bottle |
Capacity (Metric) | 100 mL |
Capacity (English) | 3.4 oz. |
Diameter (Metric) Inner Neck | 16.7 mm |
Height (Metric) With Closure | 126 mm |
Material | PFA |
Graduated | Yes |
Mouth | Narrow |