Learn More
Hanna Instruments™ Chemical Test Kit
Product Code. 10504784
Change view
Click to view available options
For Use With (Application):
Acidity Determination
Ammonia Determination in Fresh Water
Ammonia Determination in Seawater
Boron Determination
Bromine Determination
Carbon Dioxide Determination
Free Chlorine LR/MR Determination
Free Chlorine MR Determination
Free and Total Chlorine Determination
Iron (Fe2+ and Fe3+) Determination
Nitrite Determination
Phosphate Determination
Sulfate Determination
Sulfite Determination
Unit Size:
Each
Pack of 3.5
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
10504784 Iron (Fe2+ and Fe3+) Determination Each
10637554 Carbon Dioxide Determination Each
11692300 Free Chlorine LR/MR Determination Each
11612310 Free and Total Chlorine Determination Each
11676770 Nitrite Determination Each
11642380 Bromine Determination Each
11642340 Boron Determination Each
11616780 Free Chlorine MR Determination Pack of 3.5
11622300 Sulfate Determination Each
11612370 Acidity Determination Each
11642370 Sulfite Determination Each
10480684 Ammonia Determination in Fresh Water Each
11692370 Ammonia Determination in Seawater Each
10634155 Phosphate Determination Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10504784 Supplier Hanna Instruments™ Supplier No. HI38040
  • ONLINE EXCLUSIVE PRICE
    2184.00 NOK
    1655.29 NOK / Pack of 1
    Save 528.71 NOK
    24% Off
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Analytes Phenanthroline
For Use With (Application) Iron (Fe2+ and Fe3+) Determination
For Use With (Equipment) Water samples
Includes Checker™ disc, 100 packets HI3834-0 iron reagent, glass vials with caps (2), and 1 (3mL) plastic pipette
No. of Tests 100
Quantity 1 Kit
Range 0.0 to 5.0mg/L
Sufficient For Medium range determination
Test Method Titration
Type Chemical Test Kit
compliance-icons compliance-icons
Product Identifier
  • Iron Reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/first aider.
Supplemental information
  • MIXTURE LIST-Contain: sodium metabisulphitsodium dithionite, 1,10-phenanthroline