Learn More
Bochem™ Chemical Scoop
Product Code. 10653161
Change view
Click to view available options
Capacity (Metric):
120 mL
1500 mL
Length (Metric):
190 mm
350 mm
Size:
90 mm
120 mm
Unit Size:
Each
Product Code. Capacity (Metric) Length (Metric) Size unitSize
10653161 1500 mL 350 mm 120 mm Each
10569271 120 mL 190 mm 90 mm Each
2 options
Product Code. 10653161
Brand: Bochem™ 8803
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ Chemical Scoop has a round barrel.
  • Round barrel
  • High sides
  • 18/10 stainless steel
Specifications
Capacity (Metric) 1500 mL
Product Type Chemical Scoop
Size 120 mm
Material Stainless Steel (18/10)
Length (Metric) 350 mm