Learn More
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits
Product Code. 11735458
Change view
Click to view available options
Range:
6.5 to 8.4 pH, 0 to 1 mg/L Cl2
6.5 to 8.4 pH, 0 to 1 mg/L Cu
6.5 to 8.4 pH, 0 to 4 mg/L Cl2
6.5 to 8.4 pH, 0 to 5 mg/L Br
6.5 to 8.4 pH, 0.1 to 2 mg/L Cl2
Description:
pH Value
Copper
pH Value
Bromine
pH Value
Chlorine
pH value
Chlorine
Unit Size:
Each
Product Code. Range Description unitSize
11735458 6.5 to 8.4 pH, 0 to 1 mg/L Cl2 pH Value
Chlorine
Each
10631613 6.5 to 8.4 pH, 0.1 to 2 mg/L Cl2 pH Value
Chlorine
Each
10739355 6.5 to 8.4 pH, 0 to 1 mg/L Cu pH Value
Copper
Each
10193202 6.5 to 8.4 pH, 0 to 1 mg/L Cl2 pH Value
Chlorine
Each
10739104 6.5 to 8.4 pH, 0 to 4 mg/L Cl2 pH value
Chlorine
Each
10461045 6.5 to 8.4 pH, 0 to 5 mg/L Br pH Value
Bromine
Each
6 options
Product Code. 11735458
Brand: Lovibond™ 147015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
  • Easy operation and exact reagent dosing
  • Tablet reagents with a minimm guaranteed shelf life of 5/10 years
  • Rotate continuous gradient color wheel to obtain a color match with the reacted sample
  • Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
Description pH Value
Chlorine
Includes Checkit Discs, cells, stirring rod and Aqualytic tablet reagents (for 30 Tests) for the desired test
No. of Tests 30
Range 6.5 to 8.4 pH, 0 to 1 mg/L Cl2
Sufficient For 30 Tests
Test Method Colorimetric
Type 2 in 1 Test Kit