Learn More
Lovibond™ Checkit™ Comparator Single Parameter Test Kit
Click to view available options
Range:
0 to 1mg/L
4 to 10pH
5.2 to 6.8mg/L
6 to 7.6mg/L
Analytes:
Ammonia
Copper
pH
Packungsgröße:
Each
Beschreibung
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
- Compact and lightweight
- Provides the basis for a comprehensive and easy-to-use analysis system
- Comparator Discs contain a continuous color scale for exact color matching between standard and sample
Spezifikation
Spezifikation
| Analytes | Copper |
| For Use With (Application) | For Both Mobile and Static Water Analysis |
| Includes | Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions |
| Quantity | 1 Each |
| Range | 0 to 1mg/L |
| Type | Checkit™ Comparator Single Parameter Test Kit |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur