Learn More
CHCHD10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25671-1-AP-20UL
 View more versions of this product
Product Code. 16864235
  • 1710.33 NOK / 20µL
Description
CHCHD10 encodes a mitochondrial protein that is enriched at cristae junctions in the intermembrane space. It may play a role in cristae morphology maintece or oxidative phosphorylation. Mutations in this gene cause frontotemporal dementia and/or amyotrophic lateral sclerosis-2. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7 and 19.
Specifications
Specifications
CHCHD10
Polyclonal
Unconjugated
CHCHD10
C22orf16, CHCHD10, N27C7 4, Protein N27C7 4
Rabbit
Antigen Affinity Chromatography
RUO
103172, 400916
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WYQ3
CHCHD10
CHCHD10 Fusion Protein Ag22598
20 μL
Primary
Mouse, Human