missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Chapman TTC Agar (Tergitol 7 Agar)
Description
- Recommended medium for collometry of water by filtration.
- Use with sterile 1% triphenyltetrazolium chloride (TTC) solution (Ref. 06-023).
Specifications
Specifications
| Form | Solid |
| Media Type | Chapman TTC Agar (Tergitol 7 Agar) |
| Formulation | Meat Peptone 10.00 g/L, Meat Extract 5.00 g/L, Lactose 20.00 g/L, Yeast Extract 6.00 g/L, Bromothymol Blue 0.050 g/L, Tergitol™ 70.100 g/L, Agar 15.00 g/L |
| For Use With (Application) | Coulometry |
| Incubation Conditions | 36°C ± 2.0°C |
| Incubation Time | 44 ± 4 hr. |
| pH | 7.2 ±0.1 |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?