Learn More
Nabertherm™ Chamber Furnace LH 1400 Series with C540 Controller
Product Code. 17377959 Shop All Nabertherm Products
Change view
Click to view available options
Capacity (Metric):
15 L
30 L
60 L
120 L
216 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
17377959 30 L Each
17317959 15 L Each
17337969 60 L Each
17397969 120 L Each
17357979 216 L Each
5 options
Product Code. 17377959
Brand: Nabertherm™ LH0342NMNE
Only null left
Add to basket
Add to basket
Robust insulation of light refractory bricks. With a wide variety of optional equipment, these chamber furnaces can be optimally adapted to your processes.
  • Maximum Temperature: 1400°C
  • Dual shell housing with rear ventilation, provides for low shell temperatures
  • Five-sided heating for very good temperature uniformity
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Controller mounted on furnace door and removable for comfortable operation
  • Protection of bottom heating and flat stacking surface provided by embedded SiC plate in the floor
  • Multi-layered, fiber-free insulation of light refractory bricks and special backup insulation
  • Door with brick-on-brick seal, hand fitted
  • Short heating times due to high installed power
  • Side vent with bypass connection for exhaust pipe
  • Self-supporting arch for high stability and greatest possible protection against dust
  • Quick lock on door
  • Freely adjustable air slide intake in furnace floor
  • Stand included
  • Defined application within the constraints of the operating instructions
Specifications
Controller Type C540
Capacity (Metric) 30 L
Temperature (Metric) 1400°C
Dimensions (D x W x H) Interior 320 x 320 x 320 mm
Dimensions (L x W x H) Exterior 930 x 700 x 1285 mm
Weight (Metric) 200 kg