missing translation for 'onlineSavingsMsg'
Learn More

Ch-TOG Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26457-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16844915

  • 1415.13 NOK / 20µL
Estimated Shipment: 09-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a cytoskeleton-associated protein which belongs to the TOG/XMAP215 family. The N-terminal half of this protein contains a microtubule-binding domain and the C-terminal half contains a KXGS motif for binding tubulin dimers. This protein has two distinct roles in spindle formation; it protects kinetochore microtubules from depolymerization and plays an essential role in centrosomal microtubule assembly. This protein may be necessary for the proper interaction of microtubules with the cell cortex for directional cell movement. It also plays a role in translation of the myelin basic protein (MBP) mRNA by interacting with heterogeneous nuclear ribonucleoprotein (hnRNP) A2, which associates with MBP. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

Ch-TOG
Polyclonal
Unconjugated
CKAP5
ch TOG, CHTOG, CKAP5, cytoskeleton associated protein 5, MSPS, TOG, TOGp, XMAP215
Rabbit
Antigen Affinity Chromatography
RUO
9793
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14008
CKAP5
Ch-TOG Fusion Protein Ag24875
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers