Learn More
Ch-TOG Mouse anti-Human, Unconjugated, Clone: 1H5G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67631-1-IG-150UL
Description
The Ch-TOG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Ch-TOG. This antibody recognizes human, mouse, rat, pig antigen. The Ch-TOG antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Ch-TOG | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 1H5G1 | |
| Unconjugated | |
| 150 μL | |
| 9793 | |
| IgG1 |