Learn More
cFMS Receptor Inhibitor II, MedChemExpress
Product Code. 16497347
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497347 5 mg 5mg
16434638 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16497347 Supplier Medchem Express Supplier No. HY112451
MedChemExpress cFMS Receptor Inhibitor II is a CSF1R kinase inhibitor. CSF-1 is a cytokine.
cFMS Receptor Inhibitor II is a CSF1R kinase inhibitor. CSF-1 is a cytokine.
Specifications
Chemical Name or Material cFMS Receptor Inhibitor II
CAS 959860-85-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H20N4O
Quantity 5 mg
Solubility Information DMSO : 83.33 mg/mL (226.18 mM; Need ultrasonic)
SMILES O=C(C1=C(NC2=CC=C(C)C(C)=C2)C3=CC=C(C4=CC=NC=C4)C=C3N=C1)N
Molecular Weight (g/mol) 368.43
Formula Weight 368.43
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation