Learn More
Abnova™ CFL1 (Human) Recombinant Protein
Product Code. p-7164085
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16116224
Brand: Abnova™ H00001072P01.10ug
Human CFL1 full-length ORF ( AAH11005, 1 a.a. - 166 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MASGVAVSDGVIKVFNDMKVRKSSTPEEVKKRKKAVLFCLSEDKKNIILEEGKEILVGDVGQTVDDPYATFAKMLPDKDCRYALYDATYETKESKKEDLVFIFWAPESAPLKSKMIYASSKDAIKKKLTGIKHELQANCYEGVKDRCTLAEKLGGSAVISLEGKPL
Specifications
Accession Number AAH11005
Gene ID (Entrez) 1072
Name cofilin 1 (non-muscle)
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CFL
Gene Symbol CFL1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer