missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFDP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26636-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May play a role during embryogenesis.Specifications
CFDP1 | |
Polyclonal | |
Unconjugated | |
CFDP1 | |
BUCENTAUR, CFDP1, CP27, p97, SWC5, Yeti | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10428 | |
-20°C | |
Liquid |
Western Blot | |
0.83 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UEE9 | |
CFDP1 | |
CFDP1 Fusion Protein Ag24425 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |