missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Cevimeline hydrochloride hemihydrate, MedChemExpress
Description
(+)-Cevimeline hydrochloride hemihydrate ((+)-SNI-2011), a potent muscarinic receptor agonist, is a candidate therapeutic drug for xerostomia in Sjogren's syndrome.
Specifications
Specifications
| Chemical Name or Material | (+)-Cevimeline hydrochloride hemihydrate |
| Physical Form | Solid |
| Color | Light Yellow |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H17NOS.HCl.1/2H2O |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | (+)-SNI-2011(+)-AF102B hydrochloride hemihydrate |
| SMILES | C[C@H](SC1)O[C@]21CN3CCC2CC3.Cl.[0.5H2O] |
| Molecular Weight (g/mol) | 244.78 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?