missing translation for 'onlineSavingsMsg'
Learn More

(+)-Cevimeline hydrochloride hemihydrate, MedChemExpress

Product Code. 16491969
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491969 1 mg 1mg
16450658 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491969 Supplier Medchem Express Supplier No. HY76772A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (+)-Cevimeline hydrochloride hemihydrate ((+)-SNI-2011), a potent muscarinic receptor agonist, is a candidate therapeutic drug for xerostomia in Sjogren's syndrome.

(+)-Cevimeline hydrochloride hemihydrate ((+)-SNI-2011), a potent muscarinic receptor agonist, is a candidate therapeutic drug for xerostomia in Sjogren's syndrome.

Specifications

Chemical Name or Material (+)-Cevimeline hydrochloride hemihydrate
Physical Form Solid
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C10H17NOS.HCl.1/2H2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym (+)-SNI-2011(+)-AF102B hydrochloride hemihydrate
SMILES C[C@H](SC1)O[C@]21CN3CCC2CC3.Cl.[0.5H2O]
Molecular Weight (g/mol) 244.78
Formula Weight 244.78
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.