Learn More
Paragon Scientific™ Certified Viscosity Check Oil, 10W30
Product Code. p-7161819
Change view
Click to view available options
Volume (Metric):
500 mL
5 L
Unit Size:
5 L
500mL
Product Code. Volume (Metric) unitSize
16060192 500 mL 500mL
16070192 5 L 5 L
2 options
Product Code. 16060192
Brand: Paragon Scientific™ CVCO10W30
The viscosity values for the certified viscosity check oils have been tested and certified at typical blend targets. These check oils are for the validating the performance of Cold-Cranking Simulator and Kinematic Viscometers by measuring commercially available formulated engine oils
  • Certified in accordance to ISO 17025 / ISO 17034
  • Kinematic Viscosity mm2/s (cSt) in accordance with ASTM D445
  • Density g/mL in accordance with ASTM D4052
  • CCS Dynamic Viscosity mPa.s (cP) in accordance with ASTM D5293
  • Viscosity Index in accordance with ASTM D2270
  • Standard Pack Size 500mL and 5L available
  • Fully traceable to National Standards
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Specifications
Certifications/Compliance ISO 17025 / ISO 17034
Volume (Metric) 500 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
Nominal values quoted. Certified Values will vary from batch to batch.