Learn More
Certified Reference Material Fatty Acid Methyl Ester (FAME) Standard, Paragon Scientific™
Product Code. 16020040
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16020040 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16020040 Supplier Paragon Scientific Supplier No. CRMFAGO
  • ONLINE EXCLUSIVE PRICE
    3910.00 NOK
    3195.37 NOK / 250mL
    Save 714.63 NOK
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Fatty Acid Methyl Ester (FAME) Certified Reference Material for the calibration and verification of laboratory instruments used for the determination of Fatty Acid Methyl Ester (FAME) content in middle distillates - Infrared spectrometry method, EN 14078
  • Certified Reference Material for Fatty Acid Methyl Ester (FAME) Content
  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified via Round Robin
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
UN Number UN 1202
Quantity 250 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 250ML CRM-FAGO Fatty Acid Methyl Ester Standard, D
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: diesel
Nominal values quoted. Certified Values will vary from batch to batch.