Learn More
Certified Reference Material Density Standard, Gasoline, UKAS ISO 17034 Certified 250mL (nominal value: 0.73632 g/mL at 15°C), Paragon Scientific™
Product Code. 15431850
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. 15431850
Brand: Paragon Scientific CRMUDEGA
Density Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Density according to ASTM D4052. Alternative test methods are IP 365 and ISO 12185.
  • Fully certified in accordance with ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
  • Low levels of uncertainty achieves maximum accuracy of data
  • All data generated exclusively by UKAS ISO 17025 accredited laboratories
  • Fully traceable to international standards
  • Supplied in tamper evident UN compliant security pack
  • 12 month shelf life
  • Available from stock for immediate dispatch
  • Manufactured in the United Kingdom
Specifications
UN Number UN 1203
Quantity 250 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Certified Reference Material Density Standard, Gasoline
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : Gasoline : low boiling point naphtha - unspecified
Nominal values quoted. Certified Values will vary from batch to batch.