Learn More
Certified Reference Material Density Standard, Diesel, UKAS ISO 17034 Certified 250mL (nominal value: 0.84238 g/mL at 15°C), Paragon Scientific™
Product Code. 15441850
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. Quantity unitSize
15441850 250 mL 250mL
1 options
Product Code. 15441850
Brand: Paragon Scientific CRMUDEGO
Density Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Density according to ASTM D4052. Alternative test methods are IP 365 and ISO 12185.
  • Fully certified in accordance with ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
  • Low levels of uncertainty achieves maximum accuracy of data
  • All data generated exclusively by UKAS ISO 17025 accredited laboratories
  • Fully traceable to international standards
  • Supplied in tamper evident UN compliant security pack
  • 12 month shelf life
  • Available from stock for immediate dispatch
  • Manufactured in the United Kingdom
Specifications
UN Number UN 1202
Quantity 250 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • CRMU-DEGO
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
Nominal values quoted. Certified Values will vary from batch to batch.