missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CERK Recombinant Protein
Brand: Abnova™ H00064781-P01.10ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH04278 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
48.2 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MQRGPASGDPREGPPRQRREGCWGLHTCLITAARLRQSGWEHDLGVCSGSTDDKLWKLAVCKALASLLLLKCQIPMLYIDAKCLTQPGLGAVRRKLAIRGGGAGPGLPQDSSEAGPEVCTRGPDLSLSFLHTEFSIFIEHLVLLSQSSVNCVSDVCLLPRSHDGSLVSSAARGLRRPEDSSRKAFLPRSPGHPSIVYYVLV | |
DKFZp434E0211/FLJ21430/FLJ23239/KIAA1646/LK4/MGC131878/dA59H18.2/dA59H18.3/hCERK | |
CERK | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
64781 | |
CERK (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CERK | |
Human | |
Recombinant | |
Solution |