missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) Nitrate Hexahydrate, Supelco™

Product Code. 15824220
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15824220 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15824220 Supplier Supelco Supplier No. 1022710100
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extra pure

TRUSTED_SUSTAINABILITY

Specifications

CAS 10294-41-4
Quantity 100 g
Molecular Formula Ce(NO3)3 · 6 H2O
MDL Number MFCD00149631
Synonym Cerium(III) nitrate hexahydrate, Cerium(III) nitrate hexahydrate
Molecular Weight (g/mol) 434.22
Grade Research
Chemical Name or Material Cerium(III) Nitrate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.