Learn More
Cerium(III) 2-ethylhexanoate, 49% in 2-ethylhexanoic acid, Ce 12%
Product Code. 11338418
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338418 2 kg 2kg
11328418 100 g 100g
11318418 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11338418 Supplier Thermo Scientific Alfa Aesar Supplier No. 040451.A3
 Restricted Item
In Stock
Add to basket
CAS: 56797-01-4 | C24H45CeO6 | 569.73 g/mol
Used as catalyst, paint dryer, plastic Additive. As the the precursor in preparation of inorganic nanoparticles in oil-in-water microemulsions. The Toluene-based precursor solution was composed by mixing gadolinium (III) 2-ethylhexanoate and cerium (III) 2-ethylhexanoate to achieve the right stoichiometry. For the ceria/zirconia precursor, cerium(III) 2-ethylhexanoate and zirconium(IV) 2-ethylhexanoate were mixed to result in a metal molar ratio Ce/Zr of 1:1 and diluted with xylene to a total metal. Thin films of CeO 2 were prepared by a photochemical method using thin amorphous films of cerium(III) 2-ethylhexanoate as the precursor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56797-01-4
Molecular Formula C24H45CeO6
Molecular Weight (g/mol) 569.73
MDL Number MFCD00014004
InChI Key GGVUYAXGAOIFIC-UHFFFAOYNA-K
Synonym cerium iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, cerium salt, cerium tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, cerium salt 1:?, hexanoic acid, 2-ethyl-, cerium 3+ salt 3:1, hexanoic acid, 2-ethyl-, cerium 3+ salt, cerium 2-ethylhexanoate, cerous 2-ethylhexanoate, 2-ethylhexanoic acid, cerium salt, cerium hex-cem
PubChem CID 62122
IUPAC Name cerium(3+);2-ethylhexanoate
SMILES [Ce+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Specifications
Density 1.08 g/mL
Flash Point >110°C (230°F)
Quantity 2 kg
Solubility Information Insoluble in water.
Formula Weight 569.74
Concentration or Composition (by Analyte or Components) 49% in 2-ethylhexanoic acid, Ce 12%
Physical Form Viscous Liquid
Chemical Name or Material Cerium(III) 2-ethylhexanoate