Learn More
Ceritinib, MedChemExpress
Product Code. p-200098670
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
200mg
2g
500mg
50mg
5mg
Product Code. Quantity unitSize
16481186 200 mg 200mg
16471776 500 mg 500mg
16471539 1 g 1g
16442807 50 mg 50mg
16439277 5 mg 5mg
16436866 100 mg 100mg
16426728 10 mg 10mg
16420256 2 g 2g
8 options
Product Code. 16481186
Brand: Medchem Express HY15656
MedChemExpress Ceritinib (LDK378) is a selective, orally bioavailable, and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib (LDK378) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib (LDK378) shows great antitumor potency.
Ceritinib (LDK378) is a selective, orally bioavailable, and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib (LDK378) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib (LDK378) shows great antitumor potency.
Specifications
Chemical Name or Material Ceritinib
CAS 1032900-25-6
Color White
Purity Grade Notes Research
Molecular Formula C28H36ClN5O3S
Quantity 200 mg
Synonym LDK378
Solubility Information DMSO : 5.6 mg/mL (10.03 mM; Need ultrasonic) ∣Ethanol : ≥ 3.33 mg/mL (5.97 mM)
SMILES CC(C)OC1=CC(C2CCNCC2)=C(C)C=C1NC3=NC=C(Cl)C(NC4=CC=CC=C4S(=O)(C(C)C)=O)=N3
Molecular Weight (g/mol) 558.14
Formula Weight 558.14
Percent Purity 97.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease