Learn More
Cerebellin, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 94071-26-8 |
| Color | White |
| Molecular Formula | C69H113N23O23 |
| MDL Number | MFCD00167553 |
| Quantity | 1 mg |
| Synonym | cerebellin, cerebellin rat, cerebellin rat 9ci, nh2-ser-gly-ser-ala-lys-val-ala-phe-ser-ala-ile-arg-ser-thr-asn-his-oh |
| InChI Key | ADHFFUOAOLWHGU-JPDUFPOXSA-N |
| SMILES | CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@H](C)NC(=O)[C@@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)CNC(=O)[C@@H](N)CO)C(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CO)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC1=CN=CN1)C(O)=O |
| IUPAC Name | (2S)-2-[(2S)-2-[(2S,3R)-2-[(2S)-2-[(2S)-2-[(2S,3S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-6-amino-2-[(2S)-2-[(2S)-2-{2-[(2S)-2-amino-3-hydroxypropanamido]acetamido}-3-hydroxypropanamido]propanamido]hexanamido]-3-methylbutanamido]propanamido]-3-phenylpropanamido]-3-hydroxypropanamido]propanamido]-3-methylpentanamido]-5-carbamimidamidopentanamido]-3-hydroxypropanamido]-3-hydroxybutanamido]-3-carbamoylpropanamido]-3-(1H-imidazol-5-yl)propanoic acid |
| Molecular Weight (g/mol) | 1632.80 |
| Show More |