missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin Monohydrate USP MP Biomedicals

Product Code. 11427891 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 11427891

Brand: MP Biomedicals 0215058510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 23325-78-2
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.40
MDL Number MFCD00167148,MFCD00056877
InChI Key AVGYWQBCYZHHPN-CYJZLJNKSA-N
Synonym 7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid, Cefalexin monohydrate
PubChem CID 62921
ChEBI CHEBI:3535
IUPAC Name (6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
SMILES O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O

Specifications

Type Cephalexin Monohydrate
Physical Form Crystalline Powder
Color Beige to White
Quantity 10 g
Formula Weight 365.4
Grade BP/USP
Assay ≥95%
Chemical Name or Material Cephalexin Monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.