missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin monohydrate, MedChemExpress

Product Code. 16430818
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16430818

Brand: Medchem Express HYB0200B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cephalexin (Cefalexin) monohydrate is a potent, orally active new semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum.

Cephalexin (Cefalexin) monohydrate has antibacterial activity against a wide variety of gram-positive and gram-negative bacteria. Cephalexin (Cefalexin) monohydrate targets penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. Cephalexin (Cefalexin) monohydrate is used for the research of pneumonia, strep throat, and bacterial endocarditis, et al.

Specifications

Chemical Name or Material Cephalexin monohydrate
Physical Form Solid
CAS 23325-78-2
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C16H19N3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym Cefalexin hydrate Cephacillin hydrate
Solubility Information DMSO : 6.67 mg/mL (18.25 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.47 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(C)CS[C@]2([H])[C@H](NC([C@H](N)C3=CC=CC=C3)=O)C1=O)O.O
Molecular Weight (g/mol) 365.4
Formula Weight 365.4
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.