missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CEP70 plays a role in the organization of both preexisting and nascent microtubules in interphase cells. During mitosis, it is required for the organization and orientation of the mitotic spindle.
Specifications
Specifications
| Antigen | CEP70 |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CEP70 |
| Gene Accession No. | Q5PQQ9, Q6IQY5, Q8NHQ1 |
| Gene Alias | BITE, centrosomal protein 70kDa, Centrosomal protein of 70 kDa, CEP70, p10 binding protein |
| Gene Symbols | Cep70 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?