Learn More
CEP192 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Product Code. 17211416 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17211416 20 μL 20µL
17201416 150 μL 150µL
2 options
Product Code. 17211416
Brand: Proteintech 287001AP20UL
Rabbit Polyclonal Antibody
The CEP192 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CEP192. This antibody recognizes human antigen. The CEP192 antibody has been validated for the following applications: IF, WB,ELISA analysis.
Specifications
Antigen CEP192
Applications Western Blot, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 55125
Target Species Human
Form Liquid