Learn More
CEP192 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Product Code. 17201416 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17201416 150 μL 150µL
17211416 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17201416 Supplier Proteintech Supplier No. 287001AP150UL
Rabbit Polyclonal Antibody
The CEP192 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CEP192. This antibody recognizes human antigen. The CEP192 antibody has been validated for the following applications: IF, WB,ELISA analysis.
Specifications
Antigen CEP192
Applications Western Blot, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 55125
Target Species Human
Form Liquid