missing translation for 'onlineSavingsMsg'
Learn More

CEP192 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18832-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888674

  • 3468.15 NOK / 150µL
Estimated Shipment: 20-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CEP192 is required for mitotic centrosome and spindle assembly. Appears to be a major regulator of pericentriolar material (PCM) recruitment, centrosome maturation, and centriole duplication.
Specifications

Specifications

CEP192
Polyclonal
Unconjugated
CEP192
centrosomal protein 192kDa, CEP192, KIAA1569, KIAA1569 protein
Rabbit
Antigen Affinity Chromatography
RUO
55125
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8TEP8
CEP192
Peptide
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers