Learn More
Labconco CentriVap Cart
Product Code. 30533428 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30533428 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30533428 Supplier Labconco Supplier No. 8544000
  • ONLINE EXCLUSIVE PRICE
    14745.00 NOK
    13053.22 NOK / Pack of 1
    Save 1691.78 NOK
    11% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Powder-coated steel cart supports CentriVap Vacuum Concentrators, CentriVap Cold Traps, and vacuum pumps during use, storage, and transportation.
  • For use with Labconco CentriVap Pro and CentriVap Standard Vacuum Concentrator Systems.
  • Ergonomic design allows for easy access to all key components of the system.
  • Shipping weight 132.0 lbs. (59.9 kg)
Specifications
Material Stainless Steel
Type Trolley Cart
Rotor System CentriVap Pro