Learn More
TPP™ Centrifuge Tubes
Description
Intended for centrifugation of suspensions such as cells, bacteria, precipitate and others. TPP™ Centrifuge Tubes are manufactured on automatic transport units ensuring quick and careful handling without unnecessary contamination risk.
- Manufactured with highest grade of raw material fulfilling requirements of USP Class VI
- White writing circle on screw cap for easy labeling
- Instrument caps cannot be over-tightened; gas and aerosol tight cap
- Graduations down to 0.1 ml / 0.5 ml respectively
- Convenient autoclavable, collapsible racks available
Specifications
Specifications
| Capacity (Metric) | 13 mL |
| Disposable | Single-use |
| Material | Polypropylene |
| Length (Metric) | 105 mm |
| Bottom Shape | Round |
| Diameter (Metric) | 17.1 mm |
| Graduated | Yes |
| Max. RCF | 15,500 x g |