Learn More
TPP Techno Plastic Products AG Centrifuge Tube
Click to view available options
Capacity (Metric):
15 mL
50 mL
Shape:
Conical
Round
Unit Size:
Pack of 160
Pack of 360
Description
- Your valuable probes are in good hands with the centrifuge tubes from TPP! Features of the TPP centrifuge tubes surpass ordinary tubes.
- Manufactured with highest grade of raw material fulfilling requirements of USP Class VI
- White writing circle, affixed on the screw cap, for easy labeling with any writing instrument
- Caps that cannot be over-tightened
- Gas and aerosol tight cap
- Graduations all the way down to 0.1 mL / 0.5 mL respectively
- Convenient racks available that are autoclavable and are collapsible for storage when not in use
Specifications
Specifications
| Color | Opaque |
| Capacity (Metric) | 15 mL |
| Disposable | Single-use |
| Material | Polypropylene |
| No. per Pack | 40 |
| Packaging | Small Pk. |
| Sterility | Sterile |
| RNase Free | Yes |
| Autoclavable | Not Autoclavable |
| Shape | Conical |
| Show More |