Learn More
OHAUS™ Centrifuge Rack
Shop All Ohaus Products
Click to view available options
Capacity:
14 x 15 mL
17 x 15 mL
175/225 mL
18 x 15 mL
18 x 4/10 mL
21 x 10 mL
250 mL
27 x 1.8/7 mL
3 x 100 mL
32 x 7 mL
38 x 1.1/1.4 mL
5 x 50 mL
500 mL
7 x 50 mL
8 x 30 mL
Description:
FA x2
FAw/R x2
FB x2 V1
RB x2
Sar x2
Vac/Sar x2
Unit Size:
Pack of 2
Description
- Adapter for centrifuges:
- Centrifuge Multi FC5916R
- Centrifuge Multi FC5916
Specifications
Specifications
| Capacity | 17 x 15 mL |
| For Use With (Equipment) | Centrifuge Multi FC5916R and Centrifuge Multi FC5916 |
| Bore Size (Metric) | Depth 17 mm |
| Description | FA x2 |