Learn More
OHAUS™ Centrifuge Rack
Product Code. 16632774 Shop All Ohaus Products
Click to view available options
Capacity:
14 x 15 mL
17 x 15 mL
175/225 mL
18 x 15 mL
18 x 4/10 mL
21 x 10 mL
250 mL
27 x 1.8/7 mL
3 x 100 mL
32 x 7 mL
38 x 1.1/1.4 mL
5 x 50 mL
500 mL
7 x 50 mL
8 x 30 mL
Description:
FA x2
FAw/R x2
FB x2 V1
RB x2
Sar x2
Vac/Sar x2
Unit Size:
Pack of 2
Product Code. 16632774
Brand: OHAUS™ 30553131
Adapter accessory for Centrifuge Multi FC5916R and Centrifuge Multi FC5916
  • Adapter for centrifuges:
    • Centrifuge Multi FC5916R
    • Centrifuge Multi FC5916
TRUSTED_SUSTAINABILITY
Specifications
Capacity 7 x 50 mL
For Use With (Equipment) Centrifuge Multi FC5916R and Centrifuge Multi FC5916
Bore Size (Metric) Depth 29 mm
Description RB x2