Learn More
Hettich™ Centrifuge Adapters for 8-place Angel Rotors
Description
- Centrifuge Adapters for 8-place Angle Rotors
- For use with ROTOFIX 32 A and ROTINA 380/380 R Centrifuges
- For Rotors no. 1418, 1721
Specifications
Specifications
| Type | Adapter |
| Tube Type | Blood Collection/Urine, Round Bottom, Conical, Microliter |
| Capacity | 4 x 1.1 to 15 mL Tubes |
| Max. Tube Diameter | 17 mm |
| Max. Tube Length | 100 mm |
| For Use With (Equipment) | ROTOFIX 32 A and ROTINA 380/380 R Centrifuges with Rotors no. 1418, 1721 |
| Vessels per Adapter | 4 |