Learn More
Hettich™ Centrifuge Adapters for 8-place Angel Rotors
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1468
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10412305
  • 375.00 NOK / Each
Estimated Shipment: 27-06-2024
Description
  • Centrifuge Adapters for 8-place Angle Rotors
  • For use with ROTOFIX 32 A and ROTINA 380/380 R Centrifuges
  • For Rotors no. 1418, 1721
Specifications
Specifications
Adapter
50 mL
107 mm
1
Round Bottom, Conical, Skirted
29 mm
ROTOFIX 32 A and ROTINA 380/380 R Centrifuges with Rotors no. 1418, 1721