Learn More
Hettich™ Centrifuge Adapter for Hettich Angle Rotors
Product Code. 10502692
Click to view available options
Capacity:
25 mL
4 x 0.5 to 3 mL
50 mL
Unit Size:
Each
Product Code. 10502692
Brand: Hettich™ 1447
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • For Hettich Rotors no. 1620A, 1720, 1792, 4246, 4794, 5615
Specifications
Type Adapter
Capacity 25 mL
Max. Tube Diameter 26 mm
Max. Tube Length 100 mm
For Use With (Equipment) For Hettich Rotors no. 1620A, 1720, 1792, 4246, 4794, 5615
Vessels per Adapter 1
No. of Places 1