Learn More
Hettich™ Centrifuge Adapter for Hettich Angle Rotors
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1449
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10138721
  • 467.00 NOK / Each
Estimated Shipment: 05-07-2024
Description
  • For Hettich Rotors no. 1620A, 1720, 1792, 4246, 4794, 5615
Specifications
Specifications
Adapter
4 x 0.5 to 3 mL
60 mm
4
Pediatric, Conical, Round Bottom, Microliter
11 mm
For Hettich Rotors no. 1620A, 1720, 1792, 4246, 4794, 5615