Learn More
OHAUS™ Centrifuge Adapter
Shop All Ohaus Products
Click to view available options
Capacity:
15 x 1.5/2.0 mL
175/225 mL
250 mL
3 x 50 mL
4 x 30 mL
50 mL
6 x 15 mL
Description:
FA x2
FA x3
FB x2
MTB x2
RB x2
Bore Size (Metric):
Depth 11 mm
Depth 17 mm
Depth 25.5 mm
Depth 29 mm
Depth 29.5 mm
Depth 60 mm
Depth 60.5 mm
Depth 61 mm
Depth 61.5 mm
Unit Size:
Pack of 2
Pack of 3
Description
- Adapter for various Centrifuge Multi models
- Various capacities available
Specifications
Specifications
| Type | Adapter |
| Capacity | 4 x 30 mL |
| For Use With (Equipment) | Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V |
| Bore Size (Metric) | Depth 25.5 mm |
| Description | RB x2 |