Learn More
Motic™ Phase Centering Telescope
Centers phase rings for phase contrast applications. Motic™ Phase Centering Telescope replaces the use of an eyepiece to move the intermediate image plane to an observable point.
Brand: Motic™ 1101000300836
Additional Details : Weight : 0.80000kg
Description
- For use with Inverted AE30/AE31-EA Series Microscopes
- Centers phase ring
- Adjusts the the image plane to an observable point
- For phase contrast applications
Order Info
dimProductType
Specifications
Phase Centering Telescope | |
Phase contrast | |
AE30, AE31-EA Inverted Microscopes | |
1 Ea. |