Learn More
OHAUS™ Cent-O-Gram™ and Dial-O-Gram™ Balances
Product Code. 10478861 Shop All Ohaus Products
Change view
Click to view available options
Calibration:
1 x 0.01g (front beam), 10 x 1 (second beam), 100
Depth (English) Pan:
0.27 in.
Depth (Metric) Pan:
0.7 cm
Model:
311-00
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Calibration Depth (English) Pan Depth (Metric) Pan Model Unit Size
10478861 1 x 0.01g (front beam), 10 x 1 (second beam), 100 0.27 in. 0.7 cm 311-00 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10478861 Supplier OHAUS™ Supplier No. 80000010
  • ONLINE EXCLUSIVE PRICE
    4345.00 NOK
    3252.33 NOK / Pack of 1
    Save 1092.67 NOK
    25% Off
View all promotions
In Stock
Support centigram accuracy and encourage student precision. OHAUS Cent-O-Gram and Dial-O-Gram Balance comes with three-point base, special floating bearing principle, and beam design which eliminates need for level adjustment.
  • Quick zeroing by means of a zero-adjust knob at beam end
  • Built-in specific gravity platform recessed in the base
  • Stainless-steel weighing pans and bearing covers prevent rust and corrosion
  • Precision-ground knife edges and self-aligning agate bearings provide maximum sensitivity and reproducibility
  • Notched beams provide positive positioning of poise
  • Magnetic damping slows oscillation
Order Info
The Dial-O-Gram (S40027) model replaces the two front beams with a vernier dial for faster reading results.
Warranty
Five years
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer Pan 3.5 in.
Diameter (Metric) Outer Pan 9 cm
Depth (English) 7 in.
Calibration 1 x 0.01g (front beam), 10 x 1 (second beam), 100
Depth (English) Pan 0.27 in.
Depth (Metric) 17.78 cm
Depth (Metric) Pan 0.7 cm
Height (English) 9 in.
Height (Metric) 22.86 cm
Model 311-00
Material Stainless Steel
Width (English) 14 in.
Width (Metric) 35.56 cm
Warranty 5 Years
Capacity 0.6 lb., 300 g
Display Type Analog
Type Cent-O-Gram™ Balance