missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16751-1-AP-150UL
This item is not returnable.
View return policy
Description
The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPN is an additional factor required for centromere assembly.Specifications
| CENPN | |
| Polyclonal | |
| Unconjugated | |
| CENPN | |
| BM039, C16orf60, CENP N, CENPN, centromere protein N, ICEN32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361416, 55839, 72155 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5U2W4, Q96H22, Q9CZW2 | |
| Cenpn | |
| CENPN Fusion Protein Ag10219 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction