Learn More
CENPF Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 17659636 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17659636 150 μL 150µL
17669636 20 μL 20µL
2 options
Product Code. 17659636
Brand: Proteintech 285681AP150UL
Rabbit Polyclonal Antibody
CENPF Polyclonal antibody apecifically detetects CENPF antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ELISA.
Specifications
Antigen CENPF
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 700 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. NM_016343
Gene Symbols CENPF
Host Species Rabbit
Immunogen CENPF fusion protein Ag29665
Purification Method Antigen affinity purification
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 1063
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG