missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEND1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13280-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a neuron-specific protein. The similar protein in pig enhances neuroblastoma cell differentiation in vitro and may be involved in neuronal differentiation in vivo. Multiple pseudogenes have been reported for this gene.Specifications
CEND1 | |
Polyclonal | |
Unconjugated | |
CEND1 | |
BM88, BM88 antigen, CEND1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361675, 51286, 57754 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5FVI4, Q8N111, Q9JKC6 | |
Cend1 | |
CEND1 Fusion Protein Ag4024 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |