Learn More
Sartorius Cellulose Extraction Thimbles
Product Code. p-200074107 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric) Inside:
19 mm
22 mm
25 mm
26 mm
28 mm
30 mm
33 mm
35 mm
40 mm
43 mm
Length (Metric):
60 mm
70 mm
80 mm
90 mm
94 mm
100 mm
118 mm
123 mm
130 mm
150 mm
205 mm
Wall Thickness:
1.3 mm
1.5 mm
1.7 mm
1.8 mm
Unit Size:
Pack of 25
Product Code. Diameter (Metric) Inside Length (Metric) Wall Thickness unitSize
30052893 19 mm 90 mm 1.3 mm Pack of 25
30052914 22 mm 80 mm 1.3 mm Pack of 25
30052895 22 mm 100 mm 1.3 mm Pack of 25
30052920 25 mm 60 mm 1.3 mm Pack of 25
30052910 25 mm 70 mm 1.3 mm Pack of 25
30052897 25 mm 80 mm 1.3 mm Pack of 25
30052924 25 mm 100 mm 1.3 mm Pack of 25
30052921 26 mm 60 mm 1.5 mm Pack of 25
30052899 28 mm 60 mm 1.3 mm Pack of 25
30052913 28 mm 80 mm 1.3 mm Pack of 25
30052894 28 mm 100 mm 1.3 mm Pack of 25
30052922 30 mm 80 mm 1.3 mm Pack of 25
30052904 30 mm 100 mm 1.3 mm Pack of 25
30052919 33 mm 60 mm 1.3 mm Pack of 25
30052912 33 mm 80 mm 1.3 mm Pack of 25
30052909 33 mm 90 mm 1.3 mm Pack of 25
30052892 33 mm 94 mm 1.3 mm Pack of 25
30052915 33 mm 100 mm 1.3 mm Pack of 25
30052917 33 mm 118 mm 1.3 mm Pack of 25
30052896 33 mm 130 mm 1.5 mm Pack of 25
30052898 33 mm 205 mm 1.3 mm Pack of 25
30052907 35 mm 150 mm 1.7 mm Pack of 25
30052902 40 mm 100 mm 1.7 mm Pack of 25
30052908 40 mm 123 mm 1.7 mm Pack of 25
30052900 40 mm 150 mm 1.8 mm Pack of 25
30052923 43 mm 123 mm 1.5 mm Pack of 25
26 options
Product Code. 30052893
Brand: Sartorius FT1201019090
Only null left
Add to basket
Add to basket
Extraction thimbles been designed to be compatible with the Soxhlet apparatus, making them ideal for a variety of applications. Designed for versatility, the thimbles can be used with any extractor, including models like the Tectator, to facilitate efficient and high-yield extraction procedures.
  • Composed of high-purity cellulose for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inside 19 mm
Material Cellulose
Wall Thickness 1.3 mm
Grade C300
Length (Metric) 90 mm
Product Type Extraction Thimbles