Learn More
- Home
- Products
- Extraction Systems
- Extraction Thimbles
- Sartorius Cellulose Extraction Thimbles
Sartorius Cellulose Extraction Thimbles
Product Code. 30052910 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric) Inside:
19 mm
22 mm
25 mm
26 mm
28 mm
30 mm
33 mm
35 mm
40 mm
43 mm
Length (Metric):
60 mm
70 mm
80 mm
90 mm
94 mm
100 mm
118 mm
123 mm
130 mm
150 mm
205 mm
Wall Thickness:
1.3 mm
1.5 mm
1.7 mm
1.8 mm
Unit Size:
Pack of 25
| Product Code. | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | unitSize |
|---|---|---|---|---|
| 30052910 | 25 mm | 70 mm | 1.3 mm | Pack of 25 |
| 30052893 | 19 mm | 90 mm | 1.3 mm | Pack of 25 |
| 30052914 | 22 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052895 | 22 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052920 | 25 mm | 60 mm | 1.3 mm | Pack of 25 |
| 30052897 | 25 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052924 | 25 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052921 | 26 mm | 60 mm | 1.5 mm | Pack of 25 |
| 30052899 | 28 mm | 60 mm | 1.3 mm | Pack of 25 |
| 30052913 | 28 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052894 | 28 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052922 | 30 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052904 | 30 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052919 | 33 mm | 60 mm | 1.3 mm | Pack of 25 |
| 30052912 | 33 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052909 | 33 mm | 90 mm | 1.3 mm | Pack of 25 |
| 30052892 | 33 mm | 94 mm | 1.3 mm | Pack of 25 |
| 30052915 | 33 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052917 | 33 mm | 118 mm | 1.3 mm | Pack of 25 |
| 30052896 | 33 mm | 130 mm | 1.5 mm | Pack of 25 |
| 30052898 | 33 mm | 205 mm | 1.3 mm | Pack of 25 |
| 30052907 | 35 mm | 150 mm | 1.7 mm | Pack of 25 |
| 30052902 | 40 mm | 100 mm | 1.7 mm | Pack of 25 |
| 30052908 | 40 mm | 123 mm | 1.7 mm | Pack of 25 |
| 30052900 | 40 mm | 150 mm | 1.8 mm | Pack of 25 |
| 30052923 | 43 mm | 123 mm | 1.5 mm | Pack of 25 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Composed of high-purity cellulose for uncontaminated results
- Engineered for superior mechanical stability to withstand rigorous use
- Designed for a precise fit across a diverse range of extractors
- Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
Specifications
Specifications
| Diameter (Metric) Inside | 25 mm |
| Material | Cellulose |
| Wall Thickness | 1.3 mm |
| Grade | C300 |
| Length (Metric) | 70 mm |
| Product Type | Extraction Thimbles |